This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03751485_s1
See other COX10 GE Assays ›
Gene Symbol:
cox10
Gene Name:
cytochrome c oxidase assembly factor heme A:farnesyltransferase COX10
Gene Aliases:
-
Chromosome Location:
Chr.10: 39495562 - 39509373 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03751485_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cox10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001126581.1 NP_001120053.1 1155 104
GenBank mRNA BC158428.1 - 1155 104
CU075744.1 - 1129 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0