Assay Details
Gene Symbol: rmdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129926.1 | NP_001123398.1 | 4 - 5 | 716 | 66 | |
XM_012968076.2 | XP_012823530.1 | 4 - 5 | 531 | 66 | ||
GenBank mRNA | BC167344.1 | - | 4 - 5 | 716 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
