This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03738436_g1
See other HAL.1 GE Assays ›
Gene Symbol:
hal.1
Gene Name:
histidine ammonia-lyase, gene 1
Gene Aliases:
HAL1, hal, his, hstd, xhal
Chromosome Location:
Chr.3: 47694247 - 47709112 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03738436_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hal.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127093.1 NP_001120565.1 5 - 6 514 89
XM_012958610.2 XP_012814064.1 4 - 5 556 89
GenBank mRNA BC161526.1 - 5 - 6 514 89
BC166298.1 - 5 - 6 516 89
BC167682.1 - 4 - 5 495 89
CU075794.1 - 5 - 6 511 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

lyase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0