Assay Details
Gene Symbol: dner | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122791.1 | NP_001116263.1 | 543 | 63 | ||
GenBank mRNA | BC121488.1 | - | 556 | 63 | ||
BC135484.1 | - | 543 | 63 | |||
BC161016.1 | - | 543 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
