Assay Details
Gene Symbol: mea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126558.2 | NP_001120030.1 | 303 | 61 | ||
GenBank mRNA | DT452732.1 | - | 374 | 61 | ||
EL637335.1 | - | 357 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
