This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03732370_m1
See other UBA3 GE Assays ›
Gene Symbol:
uba3
Gene Name:
ubiquitin like modifier activating enzyme 3
Gene Aliases:
huba3, ube1c
Chromosome Location:
Chr.4: 131397220 - 131420380 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03732370_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: uba3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122726.1 NP_001116198.1 3 - 4 259 70
XM_012960411.2 XP_012815865.1 4 - 5 342 70
XM_012960412.1 XP_012815866.1 3 - 4 299 70
XM_012960413.1 XP_012815867.1 4 - 5 352 70
XM_018092877.1 XP_017948366.1 3 - 4 292 70
GenBank mRNA BC081344.1 - 4 - 5 269 70
BC160449.1 - 3 - 4 259 70
BC169160.1 - 3 - 4 324 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0