This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03732119_m1
See other ARL6IP4 GE Assays ›
Gene Symbol:
arl6ip4
Gene Name:
ADP ribosylation factor like GTPase 6 interacting protein 4
Gene Aliases:
sr-25, srp25
Chromosome Location:
Chr.1: 152735828 - 152743393 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03732119_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: arl6ip4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114257.1 NP_001107729.1 4 - 5 725 88
XM_012954493.2 XP_012809947.1 4 - 5 1214 88
GenBank mRNA BC158419.1 - 4 - 5 725 88
BC161359.1 - 4 - 5 725 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0