This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03730308_m1
See other FRS3 GE Assays ›
Gene Symbol:
frs3
Gene Name:
fibroblast growth factor receptor substrate 3
Gene Aliases:
frs2b, frs2beta, snt-2, snt2
Chromosome Location:
Chr.2: 62516963 - 62538622 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03730308_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: frs3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113669.1 NP_001107141.1 6 - 7 796 65
XM_012960231.2 XP_012815685.1 6 - 7 937 65
XM_012960256.2 XP_012815710.1 6 - 7 909 65
GenBank mRNA BC157660.1 - 6 - 7 796 65
BC171317.1 - 5 - 6 650 65
BC171319.1 - 5 - 6 650 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0