This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03728918_m1
See other GRAP GE Assays ›
Gene Symbol:
grap
Gene Name:
GRB2 related adaptor protein
Gene Aliases:
-
Chromosome Location:
Chr.9: 12776564 - 12812983 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03728918_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: grap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001345939.1 NP_001332868.1 4 - 5 581 73
GenBank mRNA EL668360.1 - 2 - 3 606 73
EL716670.1 - 4 - 5 581 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0