Assay Details
Gene Symbol: cabp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079086.1 | NP_001072554.1 | 3 - 4 | 459 | 86 | |
XM_012966747.2 | XP_012822201.1 | 3 - 4 | 497 | 86 | ||
XM_018095440.1 | XP_017950929.1 | 3 - 4 | 498 | 86 | ||
XM_018095441.1 | XP_017950930.1 | 3 - 4 | 500 | 86 | ||
GenBank mRNA | BC122070.1 | - | 3 - 4 | 459 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
