This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03719328_m1
See other RPS14 GE Assays ›
Gene Symbol:
rps14
Gene Name:
ribosomal protein S14
Gene Aliases:
-
Chromosome Location:
Chr.3: 6060081 - 6065597 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03719328_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rps14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016722.3 NP_001016722.1 4 - 5 570 74
XM_012959040.2 XP_012814494.1 4 - 5 484 74
XM_012959041.1 XP_012814495.1 4 - 5 538 74
XM_018092094.1 XP_017947583.1 4 - 5 524 74
GenBank mRNA BC135232.1 - 4 - 5 570 74
CR760065.2 - 4 - 5 411 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0