Assay Details
Gene Symbol: nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016573.3 | NP_001016573.1 | 5 - 6 | 557 | 71 | |
XM_012968424.2 | XP_012823878.1 | 6 - 7 | 678 | 71 | ||
XM_012968425.2 | XP_012823879.1 | 6 - 7 | 660 | 71 | ||
XM_018096258.1 | XP_017951747.1 | 6 - 7 | 651 | 71 | ||
GenBank mRNA | BC122009.1 | - | 5 - 6 | 557 | 71 | |
CR760654.2 | - | 6 - 7 | 594 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
