This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03718805_m1
See other TENT2 GE Assays ›
Gene Symbol:
tent2
Gene Name:
terminal nucleotidyltransferase 2
Gene Aliases:
gld-2, gld2, papd4, papd4-a, papd4-b, xgld2
Chromosome Location:
Chr.1: 163796875 - 163819364 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03718805_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tent2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079447.1 NP_001072915.1 10 - 11 1234 97
GenBank mRNA BC118909.1 - 10 - 11 1234 97
BC136215.1 - 11 - 12 1357 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

nucleotidyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0