This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03711422_g1
See other BIRC5.1 GE Assays ›
Gene Symbol:
birc5.1 LOC101732421
Gene Name:
baculoviral IAP repeat-containing 5, gene 1
baculoviral IAP repeat-containing protein 5.1-like
Gene Aliases:
BIR1, Su2, Survivin, Survivin1, XBIR1, api4, birc5.1-a, birc5.1-b, epr-1, svv1, xSurvivin1, xsurv
-
Chromosome Location:
Chr.2: 141207363 - 141210794 on Build UCB_Xtro_10.0
Chromosome Location:
Chr.Un|NW_016688256: 187 - 1331 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
-
Assay ID Xt03711422_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: birc5.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044454.1 NP_001037919.1 1 - 2 158 92
GenBank mRNA CR761050.2 - 1 - 2 158 92
Gene Symbol: LOC101732421
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004920927.2 XP_004920984.1 1 - 2 130 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0