Assay Details
Gene Symbol: cenpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044445.1 | NP_001037910.1 | 2 - 3 | 293 | 94 | |
XM_012961237.2 | XP_012816691.1 | 3 - 4 | 505 | 94 | ||
XM_012961238.2 | XP_012816692.1 | 3 - 4 | 440 | 94 | ||
GenBank mRNA | CR760804.2 | - | 2 - 3 | 293 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
