This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03709438_g1
See other ANAPC13.1 GE Assays ›
Gene Symbol:
anapc13.1
Gene Name:
anaphase promoting complex subunit 13, gene 1
Gene Aliases:
anapc13, swm1
Chromosome Location:
Chr.8: 15558357 - 15562657 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03709438_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: anapc13.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017240.2 NP_001017240.1 1 - 2 117 70
XM_012968478.2 XP_012823932.1 2 - 3 217 70
XM_012968479.2 XP_012823933.1 2 - 3 201 70
XM_012968480.2 XP_012823934.1 2 - 3 194 70
GenBank mRNA BC121222.1 - 146 70
CR760186.2 - 1 - 2 117 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0