This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03709198_m1
See other CAB39L GE Assays ›
Gene Symbol:
cab39l
Gene Name:
calcium binding protein 39 like
Gene Aliases:
-
Chromosome Location:
Chr.2: 146269746 - 146312017 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03709198_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cab39l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017180.2 NP_001017180.1 6 - 7 696 67
XM_012957037.1 XP_012812491.1 7 - 8 843 67
XM_012957038.2 XP_012812492.1 8 - 9 1085 67
XM_012957040.2 XP_012812494.1 7 - 8 1096 67
GenBank mRNA BC127354.1 - 7 - 8 776 67
CR760674.2 - 6 - 7 696 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0