This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03707601_m1
See other RPS10 GE Assays ›
Gene Symbol:
rps10
Gene Name:
ribosomal protein S10
Gene Aliases:
-
Chromosome Location:
Chr.2: 77236566 - 77245802 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03707601_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rps10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016718.3 NP_001016718.1 3 - 4 400 72
XM_012962754.2 XP_012818208.1 4 - 5 506 72
XM_012962759.1 XP_012818213.1 4 - 5 448 72
XM_018091267.1 XP_017946756.1 4 - 5 552 72
GenBank mRNA BC156036.1 - 3 - 4 408 72
CR760085.2 - 3 - 4 402 72
CR760262.2 - 1874 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0