This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03706816_m1
See other CRCP GE Assays ›
Gene Symbol:
crcp
Gene Name:
CGRP receptor component
Gene Aliases:
rcp9
Chromosome Location:
Chr.2: 94013750 - 94029340 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03706816_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: crcp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016478.2 NP_001016478.1 3 - 4 203 68
XM_012956972.2 XP_012812426.1 3 - 4 427 68
XM_018091257.1 XP_017946746.1 3 - 4 313 68
XM_018091258.1 XP_017946747.1 3 - 4 314 68
XM_018091259.1 XP_017946748.1 3 - 4 280 68
XM_018091260.1 XP_017946749.1 3 - 4 426 68
XM_018091261.1 XP_017946750.1 3 - 4 213 68
GenBank mRNA CR760896.2 - 3 - 4 203 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_018091260.1, XM_018091261.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0