This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03706669_m1
See other GOLGA7 GE Assays ›
Gene Symbol:
golga7
Gene Name:
golgin A7
Gene Aliases:
gcp16, golga3ap1, golga7-a, golga7-b, golga7a, hspc041
Chromosome Location:
Chr.3: 107190304 - 107203373 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03706669_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: golga7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016450.3 NP_001016450.1 4 - 5 441 86
XM_012959019.2 XP_012814473.1 5 - 6 569 86
XM_012959020.2 XP_012814474.1 5 - 6 570 86
GenBank mRNA BC170747.1 - 4 - 5 440 86
BC170779.1 - 4 - 5 440 86
CR760994.2 - 4 - 5 443 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0