Assay Details
Gene Symbol: golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016450.3 | NP_001016450.1 | 2 - 3 | 279 | 78 | |
XM_012959019.2 | XP_012814473.1 | 3 - 4 | 407 | 78 | ||
XM_012959020.2 | XP_012814474.1 | 3 - 4 | 408 | 78 | ||
GenBank mRNA | BC170747.1 | - | 2 - 3 | 278 | 78 | |
BC170779.1 | - | 2 - 3 | 278 | 78 | ||
CR760994.2 | - | 2 - 3 | 281 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
