Assay Details
Gene Symbol: golga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016232.2 | NP_001016232.1 | 2 - 3 | 758 | 67 | |
GenBank mRNA | BC167364.1 | - | 2 - 3 | 640 | 67 | |
BC171031.1 | - | 2 - 3 | 704 | 67 | ||
BC171033.1 | - | 2 - 3 | 704 | 67 | ||
CR926282.2 | - | 2 - 3 | 758 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
