Assay Details
Gene Symbol: rpl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016180.2 | NP_001016180.1 | 1 - 2 | 81 | 53 | |
XM_012971761.2 | XP_012827215.1 | 2 - 3 | 186 | 53 | ||
GenBank mRNA | BC080487.1 | - | 2 - 3 | 114 | 53 | |
CR761841.2 | - | 1 - 2 | 81 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
