This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03702352_g1
See other EBNA1BP2 GE Assays ›
Gene Symbol:
ebna1bp2 ebna1bp2
Gene Name:
EBNA1 binding protein 2
EBNA1 binding protein 2
Gene Aliases:
ebp2, nobp
-
Chromosome Location:
Chr.Un|NW_016684396: 793 - 2382 on Build UCB_Xtro_10.0
Chromosome Location:
Chr.4: 75286828 - 75294863 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
-
Assay ID Xt03702352_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ebna1bp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017151.2 NP_001017151.1 4 - 5 357 90
GenBank mRNA BC159010.1 - 4 - 5 353 90
CR760915.2 - 4 - 5 357 90
Gene Symbol: ebna1bp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004913957.3 XP_004914014.1 4 - 5 425 90
GenBank mRNA CU075329.1 - 3 - 4 326 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0