This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03701447_g1
See other YPEL1 GE Assays ›
Gene Symbol:
ypel1
Gene Name:
yippee like 1
Gene Aliases:
fksg3, ypel4
Chromosome Location:
Chr.1: 145611364 - 145626928 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03701447_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ypel1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016705.2 NP_001016705.1 4 - 5 535 97
XM_012962714.2 XP_012818168.1 4 - 5 723 97
XM_018093440.1 XP_017948929.1 4 - 5 842 97
XM_018093445.1 XP_017948934.1 4 - 5 1054 97
GenBank mRNA BC122019.1 - 4 - 5 558 97
CR760137.2 - 4 - 5 535 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0