This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03700841_m1
See other UBE2V2 GE Assays ›
Gene Symbol:
ube2v2
Gene Name:
ubiquitin conjugating enzyme E2 V2
Gene Aliases:
ddvit-1, ddvit1, edaf-1, edpf-1, edpf1, mms2, uev-2, uev2
Chromosome Location:
Chr.6: 98358844 - 98365243 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03700841_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ube2v2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001016276.2 NP_001016276.1 2 - 3 174 102
XM_018094650.1 XP_017950139.1 2 - 3 375 102
GenBank mRNA CR761621.2 - 2 - 3 174 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0