This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03699576_m1
See other NUPL2 GE Assays ›
Gene Symbol:
nupl2
Gene Name:
nucleoporin like 2
Gene Aliases:
nlp-1, nlp_1
Chromosome Location:
Chr.6: 34888870 - 34900303 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03699576_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nupl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001015793.2 NP_001015793.1 5 - 6 636 108
XM_012964643.2 XP_012820097.1 6 - 7 961 108
XM_012964645.2 XP_012820099.1 6 - 7 902 108
XM_012964646.2 XP_012820100.1 6 - 7 863 108
XM_012964647.2 XP_012820101.1 6 - 7 804 108
GenBank mRNA BC089742.1 - 5 - 6 638 108
CT025225.2 - 6 - 7 728 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0