This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03695425_m1
See other MOB2.2 GE Assays ›
Gene Symbol:
mob2.2
Gene Name:
Mps one binder kinase activator-like 2, gene 2
Gene Aliases:
hcca2, p5a5
Chromosome Location:
Chr.7: 109629577 - 109650413 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03695425_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mob2.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008165.1 NP_001008166.1 5 - 6 683 87
XM_018095358.1 XP_017950847.1 6 - 7 984 87
GenBank mRNA BC082348.1 - 5 - 6 683 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

kinase activator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0