This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03694020_m1
See other SLC16A3 GE Assays ›
Gene Symbol:
slc16a3 snrnp200
Gene Name:
solute carrier family 16 member 3
small nuclear ribonucleoprotein, U5 200kDa subunit
Gene Aliases:
mct, mct3, mct4, slc16a3-a, slc16a3-b
ascc3l1
Chromosome Location:
Chr.10: 19585920 - 19600604 on Build UCB_Xtro_10.0
Chromosome Location:
Chr.3: 109129471 - 109156467 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
-
Assay ID Xt03694020_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc16a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001007911.1 NP_001007912.1 3 - 4 420 67
XM_012966459.2 XP_012821913.1 3 - 4 505 67
GenBank mRNA BC080350.1 - 3 - 4 420 67
Gene Symbol: snrnp200
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA CT485692.1 - 3431 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0