This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03691171_m1
See other PNN GE Assays ›
Gene Symbol:
pnn
Gene Name:
pinin, desmosome associated protein
Gene Aliases:
drs, drsp, mema, pinin, pnn-a, pnn-b, sdk3
Chromosome Location:
Chr.8: 84418504 - 84433252 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03691171_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pnn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005707.1 NP_001005707.1 2 - 3 268 78
XM_018096197.1 XP_017951686.1 2 - 3 247 78
GenBank mRNA BC075283.1 - 2 - 3 268 78
CT027875.2 - 2 - 3 191 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_018096197.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0