This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03685729_m1
See other CRHR1.1 GE Assays ›
Gene Symbol:
crhr1.1
Gene Name:
corticotropin releasing hormone receptor 1, gene 1
Gene Aliases:
crf-r, crf1, crfr1, crh-r1h, crhr, crhr1f
Chromosome Location:
Chr.10: 7159373 - 7200467 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03685729_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: crhr1.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001011042.1 NP_001011042.1 11 - 12 1275 92
XM_012971695.2 XP_012827149.1 11 - 12 1351 92
XM_012971696.2 XP_012827150.1 11 - 12 1418 92
XM_012971697.2 XP_012827151.1 11 - 12 1297 92
GenBank mRNA BC084149.1 - 11 - 12 1275 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0