This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03685298_m1
See other HPGDS GE Assays ›
Gene Symbol:
hpgds
Gene Name:
hematopoietic prostaglandin D synthase
Gene Aliases:
XlGSTS1, gsts, hpgds-a, hpgds-b, pgds
Chromosome Location:
Chr.1: 67341815 - 67359417 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03685298_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hpgds
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005090.1 NP_001005090.1 5 - 6 487 95
XM_012960165.2 XP_012815619.1 6 - 7 545 95
GenBank mRNA BC077016.1 - 5 - 6 487 95
BC166947.1 - 5 - 6 448 95
CR761642.2 - 5 - 6 458 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

transferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0