This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Xt03683118_m1
See other PPIG GE Assays ›
Gene Symbol:
ppig
Gene Name:
peptidylprolyl isomerase G (cyclophilin G)
Gene Aliases:
cypg
Chromosome Location:
Chr.9: 63342823 - 63356702 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID Xt03683118_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ppig
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001234.2 NP_001001234.1 6 - 7 570 78
XM_012970274.2 XP_012825728.1 7 - 8 646 78
XM_018097070.1 XP_017952559.1 6 - 7 480 78
XM_018097071.1 XP_017952560.1 7 - 8 851 78
XM_018097072.1 XP_017952561.1 5 - 6 527 78
GenBank mRNA BC067931.2 - 6 - 7 570 78
CR761489.2 - 6 - 7 496 78
CT485720.1 - 6 - 7 594 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0