Assay Details
Gene Symbol: CIPK018 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ901246.1 | - | 4 - 5 | 416 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: CIPK018 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ901246.1 | - | 4 - 5 | 416 | 113 |
Set Membership: |
![]() ![]() ![]() ![]() |