Assay Details
Gene Symbol: VARS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195378.1 | NP_001182307.1 | 18 - 19 | 2547 | 85 | |
GenBank mRNA | AK400536.1 | - | 7 - 8 | 699 | 85 | |
EU282356.1 | - | 18 - 19 | 2547 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
