This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06942409_m1
See other MRM2 GE Assays ›
Gene Symbol:
MRM2
Gene Name:
mitochondrial rRNA methyltransferase 2
Gene Aliases:
FTSJ2
Chromosome Location:
Chr.3: 1560161 - 1567351 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06942409_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001184754.1 NP_001171683.1 1 - 2 38 80
GenBank mRNA AK240599.1 - 1 - 2 45 80
AK351595.1 - 1 - 2 45 80
AK393880.1 - 1 - 2 57 80
EU694400.1 - 1 - 2 38 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001184754.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0