This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06941854_m1
See other UGT1A6 GE Assays ›
Gene Symbol:
UGT1A6
Gene Name:
UDP glucuronosyltransferase 1 family, polypeptide A6
Gene Aliases:
mpugt1A6
Chromosome Location:
Chr.15: 133968693 - 134049029 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06941854_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UGT1A6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278750.1 NP_001265679.1 4 - 5 1350 105
XM_005672293.2 XP_005672350.1 1363 105
XM_005672294.3 XP_005672351.1 1746 105
XM_013984599.2 XP_013840053.1 1463 105
XM_013984602.2 XP_013840056.1 1356 105
GenBank mRNA AB704409.1 - 4 - 5 1350 105
AK235866.1 - 4 - 5 1364 105
AK346312.1 - 4 - 5 581 105
LC032361.1 - 4 - 5 1312 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0