This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06941504_m1
See other HBP1 GE Assays ›
Gene Symbol:
HBP1
Gene Name:
HMG-box transcription factor 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 106966060 - 106993915 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06941504_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HBP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244264.1 NP_001231193.1 10 - 11 1570 102
XM_021102059.1 XP_020957718.1 1692 102
XM_021102061.1 XP_020957720.1 1616 102
XM_021102062.1 XP_020957721.1 2945 102
XM_021102063.1 XP_020957722.1 1633 102
XM_021102064.1 XP_020957723.1 1570 102
XM_021102065.1 XP_020957724.1 1668 102
XM_021102066.1 XP_020957725.1 1569 102
GenBank mRNA AK231057.1 - 10 - 11 1570 102
AK237263.1 - 1 - 2 154 102
AK390628.1 - 2 - 3 294 102
AY609948.1 - 10 - 11 1577 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0