This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06941490_m1
See other GJA4 GE Assays ›
Gene Symbol:
GJA4
Gene Name:
gap junction protein alpha 4
Gene Aliases:
Cx37
Chromosome Location:
Chr.6: 91045430 - 91048161 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06941490_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GJA4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001244224.1 NP_001231153.1 1 - 2 49 75
GenBank mRNA AK234742.1 - 1 - 2 86 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001244224.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s)

transmembrane transport

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0