This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06941401_m1
See other RGS7BP GE Assays ›
Gene Symbol:
RGS7BP
Gene Name:
regulator of G protein signaling 7 binding protein
Gene Aliases:
-
Chromosome Location:
Chr.16: 43088784 - 43192843 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06941401_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RGS7BP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243809.1 NP_001230738.1 1 - 2 771 57
XM_021076540.1 XP_020932199.1 861 57
XM_021076541.1 XP_020932200.1 861 57
XM_021076542.1 XP_020932201.1 865 57
GenBank mRNA AK348194.1 - 1 - 2 771 57
AK390035.1 - 1 - 2 857 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0