Assay Details
Gene Symbol: SRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005666020.3 | XP_005666077.1 | 911 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: SRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005666020.3 | XP_005666077.1 | 911 | 64 |
Set Membership: |
![]() ![]() ![]() |