This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06932412_m1
See other GCNT3 GE Assays ›
Gene Symbol:
GCNT3
Gene Name:
glucosaminyl (N-acetyl) transferase 3, mucin type
Gene Aliases:
-
Chromosome Location:
Chr.1: 112424212 - 112540984 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06932412_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCNT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013992917.2 XP_013848371.1 514 84
XM_021094485.1 XP_020950144.1 281 84
XM_021094496.1 XP_020950155.1 408 84
XM_021094501.1 XP_020950160.1 309 84
XM_021094505.1 XP_020950164.1 211 84
XM_021094516.1 XP_020950175.1 606 84
XM_021094519.1 XP_020950178.1 759 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_013992917.2, XM_021094485.1, XM_021094496.1, XM_021094501.1, XM_021094505.1, XM_021094516.1, XM_021094519.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Sus scrofa

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0