This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06931972_m1
See other PRDM1 GE Assays ›
Gene Symbol:
PRDM1
Gene Name:
PR/SET domain 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 72309096 - 72335474 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06931972_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRDM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001925350.4 XP_001925385.1 1229 100
XM_005659340.3 XP_005659397.1 1143 100
XM_005659341.3 XP_005659398.1 347 100
XM_005659342.3 XP_005659399.1 1119 100
XM_013992732.2 XP_013848186.1 1146 100
XM_021090447.1 XP_020946106.1 647 100
XM_021090455.1 XP_020946114.1 1238 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Sus scrofa

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0