Assay Details
Gene Symbol: ADNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003122295.6 | XP_003122343.3 | 217 | 122 | ||
XM_005660559.3 | XP_005660616.1 | 525 | 122 | |||
XM_013993971.2 | XP_013849425.1 | 383 | 122 |

More Information
Set Membership: |
![]() ![]() |
