Assay Details
Gene Symbol: CTDSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021076470.1 | XP_020932129.1 | 1001 | 72 | ||
XM_021076472.1 | XP_020932131.1 | 987 | 72 | |||
XM_021076473.1 | XP_020932132.1 | 949 | 72 | |||
XM_021076474.1 | XP_020932133.1 | 938 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
