This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06925378_m1
See other MFSD13A GE Assays ›
Gene Symbol:
MFSD13A
Gene Name:
major facilitator superfamily domain containing 13A
Gene Aliases:
-
Chromosome Location:
Chr.14: 113454507 - 113478699 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06925378_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MFSD13A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001928845.7 XP_001928880.1 991 73
XM_005671422.3 XP_005671479.1 904 73
XM_013983693.2 XP_013839147.1 1147 73
XM_013983694.2 XP_013839148.1 963 73
XM_013983695.2 XP_013839149.1 904 73
XM_021072748.1 XP_020928407.1 2609 73
XM_021072749.1 XP_020928408.1 733 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0