This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06924390_g1
See other TJAP1 GE Assays ›
Gene Symbol:
TJAP1
Gene Name:
tight junction associated protein 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 38475181 - 38501532 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06924390_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TJAP1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_021098759.1
XP_020954418.1
1321
68
XM_021098760.1
XP_020954419.1
1339
68
XM_021098762.1
XP_020954421.1
1639
68
XM_021098763.1
XP_020954422.1
2305
68
XM_021098764.1
XP_020954423.1
1022
68
XM_021098765.1
XP_020954424.1
618
68
XM_021098766.1
XP_020954425.1
1019
68
XM_021098767.1
XP_020954426.1
1149
68
XM_021098768.1
XP_020954427.1
1598
68
XM_021098769.1
XP_020954428.1
1185
68
XM_021098770.1
XP_020954429.1
1321
68
XM_021098771.1
XP_020954430.1
1451
68
XM_021098772.1
XP_020954431.1
2838
68
XM_021098773.1
XP_020954432.1
1989
68
XM_021098774.1
XP_020954433.1
3648
68
XM_021098775.1
XP_020954434.1
3645
68
XM_021098776.1
XP_020954435.1
618
68
XM_021098777.1
XP_020954436.1
1018
68
XM_021098778.1
XP_020954437.1
748
68
XM_021098779.1
XP_020954438.1
615
68
XM_021098780.1
XP_020954439.1
766
68
XM_021098781.1
XP_020954440.1
1283
68
XM_021098782.1
XP_020954441.1
1598
68
XM_021098783.1
XP_020954442.1
1281
68
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Sus scrofa
Panther Classification:
Molecular Function -
tight junction
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0