This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06921383_m1
See other IER3IP1 GE Assays ›
Gene Symbol:
IER3IP1
Gene Name:
immediate early response 3 interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 96740718 - 96763875 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06921383_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IER3IP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003353714.4 XP_003353762.1 324 137
GenBank mRNA AK232828.1 - 2 - 3 285 137
AK351638.1 - 2 - 3 253 137

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0