This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06920478_g1
See other ILK GE Assays ›
Gene Symbol:
ILK
Gene Name:
integrin linked kinase
Gene Aliases:
-
Chromosome Location:
Chr.9: 3145647 - 3152097 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06920478_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ILK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003357131.4 XP_003357179.1 361 73
XM_005656596.3 XP_005656653.1 506 73
XM_005656597.3 XP_005656654.1 503 73
XM_021062334.1 XP_020917993.1 486 73
XM_021062335.1 XP_020917994.1 497 73
XM_021062336.1 XP_020917995.1 359 73
GenBank mRNA AK232445.1 - 3 - 4 429 73
AK236668.1 - 3 - 4 359 73
AK390096.1 - 3 - 4 433 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0