This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ss06920343_m1
See other ABHD14B GE Assays ›
Gene Symbol:
ABHD14B
Gene Name:
abhydrolase domain containing 14B
Gene Aliases:
-
Chromosome Location:
Chr.13: 34100994 - 34104773 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID Ss06920343_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABHD14B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003358459.4 XP_003358507.1 351 75
XM_005669622.3 XP_005669679.1 436 75
XM_005669623.3 XP_005669680.1 370 75
XM_021068875.1 XP_020924534.1 941 75
GenBank mRNA AK232369.1 - 2 - 3 292 75
AK233194.1 - 2 - 3 292 75
AK346285.1 - 2 - 3 356 75
AK398065.1 - 2 - 3 358 75
AK398288.1 - 2 - 3 318 75
AY609928.1 - 1 - 2 350 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0